siRNA Details
Gene Symbol: Cyp2d11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104531.1 | NP_001098001.1 | 7 | 1242 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cyp2d11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104531.1 | NP_001098001.1 | 7 | 1242 |