siRNA Details
Gene Symbol: Kalrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164268.1 | NP_001157740.1 | 19 | 3241 | ||
NM_177357.3 | NP_796331.2 | 19 | 3241 | |||
GenBank mRNA | AK147314.1 | - | 19 | 3262 | ||
AK147518.1 | - | 19 | 3262 | |||
AK158544.1 | - | 10 | 1489 | |||
AK159031.1 | - | 9 | 1604 | |||
BC157950.1 | - | 19 | 3241 | 9088125 | ||
BC172101.1 | - | 19 | 3241 | 9054387 |
