siRNA Details
Gene Symbol: Zfp607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024726.2 | NP_001019897.2 | 5 | 1012 | ||
GenBank mRNA | AK028967.1 | - | 5 | 991 | ||
AK143371.1 | - | 5 | 979 | |||
AK163602.1 | - | 5 | 1012 | |||
BC042662.1 | - | 2668 | ||||
BC141161.1 | - | 5 | 979 | 9055773 |
