siRNA Details
Gene Symbol: Foxo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018789.2 | NP_061259.1 | 2 | 1659 | ||
GenBank mRNA | AB032770.1 | - | 2 | 1439 | ||
AF114260.1 | - | 2 | 1659 | |||
BC139275.1 | - | 2 | 1500 | 8862297 | ||
BC139304.1 | - | 2 | 1500 | 8862326 |
