siRNA Details
Gene Symbol: Gramd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033498.1 | NP_001028670.1 | 1, 2 | 340 | ||
GenBank mRNA | AK040545.1 | - | 2, 3 | 229 | ||
AK041631.1 | - | 1, 2 | 340 | |||
BC138948.1 | - | 1, 2 | 157 | 8861970 | ||
BC138949.1 | - | 1, 2 | 157 | 8861971 |
