siRNA Details
Gene Symbol: Arid3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017362.2 | NP_001017362.1 | 5, 6 | 972 | ||
NM_001252622.1 | NP_001239551.1 | 5, 6 | 972 | |||
GenBank mRNA | BC141237.1 | - | 4, 5 | 941 | 9055849 | |
BC145563.1 | - | 4, 5 | 941 | 9054128 |
