siRNA Details
Gene Symbol: Hebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019487.3 | NP_062360.1 | 3, 4 | 691 | ||
GenBank mRNA | AF117614.1 | - | 3, 4 | 621 | ||
AK078041.1 | - | 671 | ||||
AK078130.1 | - | 3, 4 | 691 |
