siRNA Details
Gene Symbol: Il17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019508.1 | NP_062381.1 | 3 | 576 | ||
GenBank mRNA | AF184970.1 | - | 3 | 576 | ||
AF218726.1 | - | 3 | 576 | |||
AK003506.1 | - | 3 | 634 | |||
BC002271.1 | - | 3 | 576 | 3496106 |
