siRNA Details
Gene Symbol: Astn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019514.3 | NP_062387.2 | 3 | 883 | ||
NM_207109.2 | NP_996992.1 | 3 | 883 | |||
GenBank mRNA | AK041160.1 | - | 4 | 787 | ||
AK047058.1 | - | 3 | 872 | |||
AK051472.1 | - | 3 | 884 |
