siRNA Details
Gene Symbol: Arl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019665.3 | NP_062639.3 | 2 | 470 | ||
GenBank mRNA | AB232697.1 | - | 1 | 33 | ||
AF031903.1 | - | 2 | 188 | |||
AK003778.1 | - | 2 | 298 | |||
AK152980.1 | - | 2 | 274 | |||
AK158832.1 | - | 2 | 523 | |||
AK160666.1 | - | 2 | 289 | |||
BC018497.1 | - | 2 | 290 | 3484868 |
