siRNA Details
Gene Symbol: Arl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019665.3 | NP_062639.3 | 5 | 764 | ||
GenBank mRNA | AB232697.1 | - | 4 | 327 | ||
AF031903.1 | - | 5 | 482 | |||
AK003778.1 | - | 5 | 592 | |||
AK152980.1 | - | 5 | 568 | |||
AK158832.1 | - | 5 | 817 | |||
AK160666.1 | - | 5 | 583 | |||
BC018497.1 | - | 5 | 584 | 3484868 |
