siRNA Details
Gene Symbol: Tmeff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019790.4 | NP_062764.1 | 8 | 1261 | ||
GenBank mRNA | AB017270.1 | - | 8 | 1224 | ||
AB041565.1 | - | 8 | 1266 | |||
AK029307.1 | - | 8 | 1261 | |||
AK076254.1 | - | 8 | 1261 | |||
AK135038.1 | - | 8 | 1248 | |||
BC034850.1 | - | 8 | 1237 | 1397175 |
