siRNA Details
Gene Symbol: 1110004F10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019772.2 | NP_062746.1 | 4, 5 | 399 | ||
GenBank mRNA | AB025407.1 | - | 4, 5 | 399 | ||
AK003415.1 | - | 4, 5 | 440 | |||
AK144059.1 | - | 4, 5 | 426 | |||
AK167571.1 | - | 4, 5 | 428 | |||
AK168206.1 | - | 4, 5 | 433 | |||
AK169309.1 | - | 4, 5 | 426 | |||
AK170172.1 | - | 2, 3 | 157 | |||
BC072575.1 | - | 4, 5 | 435 | 5715413 |
