siRNA Details
Gene Symbol: Nudt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019837.2 | NP_062811.1 | 1 | 309 | ||
GenBank mRNA | AF264064.1 | - | 1 | 308 | ||
BC016534.1 | - | 2 | 215 | 2615904 | ||
BC046805.1 | - | 1 | 310 | 6829170 |
Gene Symbol: Anks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC057331.1 | - | 301 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001024243, BC093618.
Gene Ontology Categories:
