siRNA Details
Gene Symbol: Pias1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019663.3 | NP_062637.2 | 9, 10 | 1171 | ||
GenBank mRNA | AF077950.1 | - | 9, 10 | 1163 | ||
AK013753.1 | - | 5, 6 | 505 | |||
AK075708.1 | - | 9, 10 | 1172 | |||
AK153871.1 | - | 9, 10 | 1176 | |||
BC051417.1 | - | 9, 10 | 1168 | 6515027 |
