siRNA Details
Gene Symbol: Vapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019806.5 | NP_062780.2 | 6 | 840 | ||
GenBank mRNA | AF115504.1 | - | 6 | 718 | ||
AK050632.1 | - | 6 | 789 | |||
AK081908.1 | - | 6 | 769 | |||
AK160925.1 | - | 6 | 683 | |||
AK172080.1 | - | 6 | 613 | |||
BC048231.1 | - | 6 | 840 | 6313869 | ||
BC057123.1 | - | 6 | 829 | 6411688 |
