siRNA Details
Gene Symbol: Ick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163780.1 | NP_001157252.1 | 10, 11 | 1799 | ||
NM_019987.2 | NP_064371.2 | 10, 11 | 1727 | |||
GenBank mRNA | AF225918.1 | - | 10, 11 | 1567 | ||
AK052178.1 | - | 3, 4 | 696 | |||
AK087484.1 | - | 10, 11 | 1651 | |||
AK133537.1 | - | 10, 11 | 1666 | |||
AK154185.1 | - | 10, 11 | 1800 | |||
AK173068.1 | - | 5, 6 | 956 | |||
BC028863.1 | - | 9, 10 | 1484 | 4224269 |
