siRNA Details
Gene Symbol: Brinp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019967.2 | NP_064351.2 | 3 | 787 | ||
GenBank mRNA | AB060589.1 | - | 3 | 787 | ||
AF202896.1 | - | 2 | 353 | |||
AK028321.1 | - | 3 | 702 | |||
AK045420.1 | - | 3 | 669 | |||
BC048789.1 | - | 3 | 755 | |||
BC079630.1 | - | 3 | 754 | 5694980 |
