siRNA Details
Gene Symbol: Pdxp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020271.3 | NP_064667.2 | 1 | 132 | ||
GenBank mRNA | AB041662.1 | - | 1 | 26 | ||
AK043228.1 | - | 1 | 93 | |||
AY366300.1 | - | 1 | 94 | |||
BC046955.1 | - | 1 | 83 | |||
BC049107.1 | - | 1 | 132 | |||
BY060098.1 | - | 1 | 102 | |||
CX236940.1 | - | 1 | 188 |
