siRNA Details
Gene Symbol: Zim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF365932.1 | - | LIT1822 Zim3 |
6 | 1451 |
Gene Symbol: Usp29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290994.1 | NP_001277923.1 | 9 | 3057 | ||
NM_021323.3 | NP_067298.2 | 8 | 2993 | |||
GenBank mRNA | AF229257.1 | - | 9 | 3057 | ||
EU233992.1 | - | 5 | 2334 |
