siRNA Details
Gene Symbol: Cldn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160096.1 | NP_001153568.1 | 3 | 628 | ||
NM_001160097.1 | NP_001153569.1 | 3 | 685 | |||
NM_001160098.1 | NP_001153570.1 | 3 | 628 | |||
NM_001160099.1 | NP_001153571.1 | 3 | 445 | |||
NM_021386.4 | NP_067361.2 | 3 | 445 | |||
NM_023878.3 | NP_076367.2 | 3 | 685 | |||
GenBank mRNA | AF124425.1 | - | 3 | 414 | ||
AK020131.1 | - | 3 | 649 | |||
BC021770.1 | - | 3 | 638 | 5040100 | ||
BC029019.1 | - | 438 | 5356256 | |||
CA481582.1 | - | 3 | 636 | |||
CK332009.1 | - | 3 | 250 | |||
CK794879.1 | - | 3 | 445 |
