siRNA Details
Gene Symbol: Cldn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160096.1 | NP_001153568.1 | 4, 5 | 795 | ||
NM_021386.4 | NP_067361.2 | 4, 5 | 612 | |||
NM_023878.3 | NP_076367.2 | 4, 5 | 852 | |||
GenBank mRNA | AF124425.1 | - | 4, 5 | 581 | ||
AI851016.1 | - | 1, 2 | 345 | |||
AK020131.1 | - | 4, 5 | 816 | |||
BC012238.1 | - | 2, 3 | 153 | |||
BC029019.1 | - | 605 | 5356256 | |||
CK332009.1 | - | 4, 5 | 417 | |||
CK794879.1 | - | 4, 5 | 612 |
