siRNA Details
Gene Symbol: Fam181b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021427.2 | NP_067402.2 | 1 | 506 | ||
GenBank mRNA | AB041569.1 | - | 1 | 466 | ||
AK043957.1 | - | 1 | 521 | |||
AK140685.1 | - | 1 | 458 | |||
AK141493.1 | - | 1 | 104 | |||
BC043329.1 | - | 1 | 455 | 5369836 |
Gene Symbol: Gm9934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047323.1 | - | 1 | 408 | ||
AK079653.1 | - | 1 | 639 |
