siRNA Details
Gene Symbol: Med12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021521.2 | NP_067496.2 | 10, 11 | 1656 | ||
GenBank mRNA | AF071310.1 | - | 9, 10 | 1406 | ||
BC057119.1 | - | 9, 10 | 1390 | |||
BC090654.1 | - | 10, 11 | 1656 | |||
BC145447.1 | - | 9, 10 | 1341 |
