siRNA Details
Gene Symbol: Ms4a4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021718.2 | NP_068364.1 | 4 | 417 | ||
GenBank mRNA | AF237913.1 | - | 4 | 386 | ||
AF354929.1 | - | 4 | 374 | |||
AK039824.1 | - | 4 | 392 | |||
AK080934.1 | - | 3 | 3443 | |||
AK088631.1 | - | 4 | 373 | |||
AK088780.1 | - | 4 | 423 | |||
AK138737.1 | - | 4 | 390 | |||
AK172495.1 | - | 4 | 390 | |||
BC028433.1 | - | 4 | 374 | 1248222 |
