siRNA Details
Gene Symbol: Tmem253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033805.3 | NP_001028977.1 | 2 | 198 | ||
GenBank mRNA | AK144313.1 | - | 2 | 199 | ||
BC147689.1 | - | 2 | 144 | 9056662 | ||
BC172129.1 | - | 2 | 144 |
