siRNA Details
Gene Symbol: Trbv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF479018.1 | - | 172 | |||
AK138971.1 | - | 241 | ||||
M16680.1 | - | 2 | 172 |

More Information
Other Species Targeted


UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trbv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF479018.1 | - | 172 | |||
AK138971.1 | - | 241 | ||||
M16680.1 | - | 2 | 172 |