siRNA Details
Gene Symbol: Mcidas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037914.3 | NP_001033003.1 | 7 | 874 | ||
GenBank mRNA | AK134107.1 | - | 7 | 875 | ||
BC150962.1 | - | 7 | 897 | 9087873 |
Gene Symbol: 6330437I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018205.1 | - | 1 | 332 |
