siRNA Details
Gene Symbol: Slc38a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037717.3 | NP_001032806.2 | 16 | 1417 | ||
GenBank mRNA | BC043932.1 | - | 9 | 744 | ||
BC157917.1 | - | 16 | 1366 | 9007345 | ||
BC157928.1 | - | 16 | 1366 | 9088101 |
