siRNA Details
Gene Symbol: Slc38a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037717.3 | NP_001032806.2 | 9 | 745 | ||
GenBank mRNA | BC043932.1 | - | 2 | 72 | ||
BC157917.1 | - | 9 | 694 | 9007345 | ||
BC157928.1 | - | 9 | 694 | 9088101 |
