siRNA Details
Gene Symbol: Gm11992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037928.3 | NP_001033017.1 | 5, 6 | 1149 | ||
GenBank mRNA | AK154909.1 | - | 5, 6 | 1150 | ||
AK160996.1 | - | 5, 6 | 1128 | |||
BC147253.1 | - | 5, 6 | 1028 | 9056290 | ||
BC147254.1 | - | 5, 6 | 1028 | 9056286 |
