siRNA Details
Gene Symbol: Zfp800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081678.1 | NP_001075147.1 | 5 | 757 | ||
GenBank mRNA | AK036691.1 | - | 6 | 1023 | ||
BC066073.1 | - | 5 | 1169 | |||
BC119236.1 | - | 5 | 757 | 8733985 | ||
BC119262.1 | - | 5 | 757 | 8734011 | ||
BC144761.1 | - | 5 | 757 | 9053309 |
