siRNA Details
Gene Symbol: Col6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167923.1 | NP_001161395.1 | 5 | 1803 | ||
GenBank mRNA | AM748256.1 | - | 5 | 1803 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Col6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167923.1 | NP_001161395.1 | 5 | 1803 | ||
GenBank mRNA | AM748256.1 | - | 5 | 1803 |