siRNA Details
Gene Symbol: Susd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163288.2 | NP_001156760.2 | 14 | 2368 | ||
GenBank mRNA | BC157977.1 | - | 14 | 2293 | 9088138 | |
BC158124.1 | - | 14 | 2345 | 9007393 |
