siRNA Details
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162500.1 | NP_001155972.1 | 8, 9 | 767 | ||
NM_022321.4 | NP_071716.3 | 8, 9 | 722 | |||
GenBank mRNA | AF312712.1 | - | 8, 9 | 680 | ||
AK028329.1 | - | 8, 9 | 723 | |||
AK037365.1 | - | 8, 9 | 695 | |||
AK040914.1 | - | 8, 9 | 653 | |||
AK079881.1 | - | 8, 9 | 648 | |||
AK088419.1 | - | 8, 9 | 672 | |||
BC011200.1 | - | 8, 9 | 676 | 4190510 |
