siRNA Details
Gene Symbol: Nip7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025391.2 | NP_079667.2 | 4 | 447 | ||
GenBank mRNA | AK003739.1 | - | 4 | 447 | ||
AK006057.1 | - | 4 | 427 | |||
AK014290.1 | - | 4 | 406 | |||
BC003972.1 | - | 4 | 434 | 3488726 | ||
BE623972.1 | - | 3 | 511 |
