siRNA Details
Gene Symbol: Tmem208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025486.2 | NP_079762.1 | 4 | 369 | ||
GenBank mRNA | AK002499.1 | - | 4 | 357 | ||
AK005662.1 | - | 4 | 353 | |||
AK131833.1 | - | 4 | 338 | |||
BC005665.1 | - | 4 | 324 | |||
BC024608.1 | - | 4 | 320 | 4242513 |
