siRNA Details
Gene Symbol: Tmem208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025486.2 | NP_079762.1 | 4, 5 | 399 | ||
GenBank mRNA | AK002499.1 | - | 4, 5 | 387 | ||
AK005662.1 | - | 4, 5 | 383 | |||
BC005665.1 | - | 4, 5 | 354 | |||
BC024608.1 | - | 4, 5 | 350 | 4242513 | ||
BE983091.1 | - | 2, 3 | 389 |
