siRNA Details
Gene Symbol: Knop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168218.1 | NP_001161690.1 | 1 | 272 | ||
NM_001168219.1 | NP_001161691.1 | 1 | 272 | |||
NM_023197.3 | NP_075686.2 | 1 | 272 | |||
GenBank mRNA | AF034580.1 | - | 2 | 439 | ||
AK006113.1 | - | 1 | 276 |
