siRNA Details
Gene Symbol: BB187676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046110.1 | - | 1 | 411 | ||
BB313560.2 | - | 1 | 398 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BB187676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046110.1 | - | 1 | 411 | ||
BB313560.2 | - | 1 | 398 |