siRNA Details
Gene Symbol: Lrrc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159609.1 | NP_001153081.1 | 6 | 894 | ||
NM_001159610.1 | NP_001153082.1 | 6 | 945 | |||
NM_001159612.1 | NP_001153084.1 | 6 | 961 | |||
NM_025657.3 | NP_079933.2 | 6 | 844 | |||
GenBank mRNA | AK003037.1 | - | 6 | 751 | ||
AK003592.1 | - | 6 | 862 | |||
AK015684.1 | - | 6 | 896 | |||
AK134971.1 | - | 6 | 803 | |||
AK169249.1 | - | 6 | 868 | |||
BC034894.1 | - | 6 | 803 | 3328402 |
