siRNA Details
Gene Symbol: Cdip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025670.4 | NP_079946.2 | 4 | 381 | ||
GenBank mRNA | AK005157.1 | - | 4 | 367 | ||
AK017480.1 | - | 4 | 364 | |||
AK031705.1 | - | 4 | 383 | |||
AK035791.1 | - | 3 | 1055 | |||
AK051560.1 | - | 4 | 379 | |||
AK149939.1 | - | 4 | 377 | |||
AK150219.1 | - | 4 | 377 | |||
AK166141.1 | - | 4 | 374 | |||
BC004063.1 | - | 4 | 380 | 3590511 |
