siRNA Details
Gene Symbol: Lix1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025681.2 | NP_079957.2 | 2 | 420 | ||
GenBank mRNA | AF351204.1 | - | 2 | 161 | ||
AK012220.1 | - | 2 | 228 | |||
AK017685.1 | - | 2 | 239 | |||
BC049574.1 | - | 2 | 434 | 6593108 | ||
BC052700.1 | - | 2 | 220 | 6416835 | ||
BC063057.1 | - | 2 | 213 | 5715784 |
