siRNA Details
Gene Symbol: Zfp541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099277.1 | NP_001092747.1 | 2 | 367 | ||
GenBank mRNA | BC157962.1 | - | 2 | 388 | 9007287 | |
DQ864732.1 | - | 2 | 367 |

More Information
Other Species Targeted

Gene Ontology Categories:
