siRNA Details
Gene Symbol: Rbm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025717.3 | NP_079993.2 | 3 | 624 | ||
GenBank mRNA | AK006447.1 | - | 3 | 607 | ||
AK142116.1 | - | 3 | 579 | |||
AK160796.1 | - | 3 | 1019 | |||
AK181241.1 | - | 1 | 31 | |||
AK182975.1 | - | 1 | 31 | |||
BC019488.1 | - | 3 | 529 | 4457936 | ||
BF782018.1 | - | 640 |
