siRNA Details
Gene Symbol: 9930111J21Rik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114679.1 | NP_001108151.1 | 2737 |
Gene Symbol: 9930111J21Rik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC066104.1 | - | 2738 |
