siRNA Details
Gene Symbol: Kcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK160694.1 | - | 247 | |||
BC059909.1 | - | 12 | 2604 |
Gene Symbol: Gm9047 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145360.1 | NP_001138832.1 | 2 | 563 | ||
GenBank mRNA | BC147635.1 | - | 2 | 563 | 9056625 | |
BC147645.1 | - | 2 | 563 | 9056635 |
