siRNA Details
Gene Symbol: Pccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025835.2 | NP_080111.1 | 2 | 293 | ||
GenBank mRNA | AF327060.1 | - | 2 | 281 | ||
AK004976.1 | - | 2 | 294 | |||
AK050366.1 | - | 2 | 299 | |||
AK134437.1 | - | 2 | 304 | |||
BC026531.1 | - | 2 | 269 | |||
BC037082.1 | - | 2 | 295 | 5098790 | ||
BC055946.1 | - | 2 | 300 | 4166700 |
