siRNA Details
Gene Symbol: Mettl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025907.3 | NP_080183.1 | 6 | 1051 | ||
GenBank mRNA | AK005448.1 | - | 6 | 1106 | ||
AK006715.1 | - | 5 | 958 | |||
AK082949.1 | - | 6 | 1108 | |||
BC030449.1 | - | 6 | 1052 | 5400506 |
