siRNA Details
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175114.3 | NP_780323.2 | 16 | 2301 | ||
GenBank mRNA | AK029123.1 | - | 18 | 2770 | ||
AK122426.1 | - | 18 | 2484 | |||
BC058971.1 | - | 16 | 2243 | 6832507 | ||
BC082807.1 | - | 3 | 267 |
